Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Bromide, Anhydrous, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog No. 18607818 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
6 x 500 g
12 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-818 25 g Amber Glass Bottle
18-607-819 100 g Amber Glass Bottle
18-607-820 500 g Amber Glass Bottle
18-607-821 6 x 500 g Amber Glass Bottle
18-607-823 12 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-818 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z104825GM
Add to Cart
Add to Cart

Z1048, 7699-45-8, ZnBr2

Spectrum™ Chemical Zinc Bromide, Anhydrous, Reagent is a salt that has no color and share many similarities with zinc chloride. This inorganic compound is very soluble in water and is used as a Lewis acid and in an aqueous solution as a transparent shield against radiation in between two glass panes. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Chemical Identifiers

CAS 7699-45-8
Molecular Formula Br2Zn
Molecular Weight (g/mol) 225.19
InChI Key VNDYJBBGRKZCSX-UHFFFAOYSA-L
IUPAC Name zinc(2+) dibromide
SMILES [Zn++].[Br-].[Br-]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 98%
Grade Reagent
Packaging Amber Glass Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.