Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc bromide, Puriss., anhydrous,≥98%, Solstice

Catalog No. 60048283 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-283 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-283 Supplier Solstice Supplier No. 21284
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Manual Blending Steps Eliminated with LabReady Blends, saved time and costs

  • By handling your blend preparation, we cut the high cost of disposing out-of-specification chemical blends while sparing your chemical inventories

  • Tested to meet isocratic and gradient HPLC requirements, as well as most LC-MS requirements

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1310-73-2 , 75-05-8 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.04, 16.40, 83.60
Density 0.969 g/cm3
Quantity 4 x 4 L
Flash Point 18.3°C
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 72.7°C
Grade HPLC
Hazard Category Danger
Hazard Statement H225+H314+H370
Precautionary Statement P280+P270+P264+P260+P243+P242+P241+P240+P233+P210
Recommended Storage Room Temperature
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.