Learn More
TA Instruments ZINC CALIBRATION MATERIAL
Supplier:  TA Instruments 900907.901
Encompass
Zinc Calibration Material (not certified): standard zinc calibration material (not certified) used for temperature calibration of DSC cells and TMA instruments.
Catalog No. NC1273295
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.