Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Carbonate, Basic, Powder, Reagent, 65%, Spectrum™ Chemical
SDP

Catalog No. 18607827 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-827 500 g Poly Bottle
18-607-825 25 g Poly Bottle
18-607-828 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-607-827 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z1050500GM
Add to Cart
Add to Cart

Z1050, 5263-02-5, 3Zn(OH)2·2ZnCO3

Spectrum™ Chemical Zinc Carbonate, Basic, Powder, Reagent

Chemical Identifiers

CAS 5263-02-5
Molecular Formula C2H6O12Zn5
Molecular Weight (g/mol) 548.96
MDL Number MFCD00011519
InChI Key UOURRHZRLGCVDA-UHFFFAOYSA-D
IUPAC Name pentazinc(2+) hexahydroxide dicarbonate
SMILES [OH-].[OH-].[OH-].[OH-].[OH-].[OH-].[Zn++].[Zn++].[Zn++].[Zn++].[Zn++].[O-]C([O-])=O.[O-]C([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 65%
Grade Reagent
Packaging Poly Bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.