Learn More
Specifications
Specifications
| Boiling Point | 732°C |
| Melting Point | 293°C |
| Color | White |
| pH | 5 |
| Physical Form | Solid |
| Quantity | 3 kg |
| Assay Percent Range | ≥97 % |
| UN Number | 2331 |
| Formula Weight | 136.3 |
| Percent Purity | ≥97% |
| Show More |
Safety and Handling
| Hazard Statement | DANGER! |
| Precautionary Statement | Emergency Overview Causes burns by all exposure routes. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Hygroscopic. Use personal protective equipment. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Immediate medical attention is required. NFPA Health:3 Flammability:1 Instability:1 |
| DOTInformation | DOT Class 8, : Corrosive |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.