Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc chloride hydrate, Puratronic™, 99.999% (metals basis)

Catalog No. AA3578209
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3578209 10 g
AA3578218 50 g
AA3578230 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA3578209 Supplier Thermo Scientific Chemicals Supplier No. 03578209
Add to Cart

CAS: 29604-34-0 | Cl2Zn | 136.28 g/mol

It is used for a source of the zinc ion. It can also be used alone or with phenols for preserving railway ties, fireproofing timber, and with ammonium chloride as a flux for soldering and etching of metals. It is used as a moderate-strength lewis acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 29604-34-0
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
MDL Number MFCD00011295
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym zinc chloride, zinc dichloride, zinc chloride zncl2, zinc butter, zinc chloride fume, zinc ii chloride, zinkchloride, zintrace, zinc chloride, anhydrous, zine dichloride
PubChem CID 5727
ChEBI CHEBI:49976

Specifications

Color White
Physical Form Crystalline Aggregates
Quantity 10 g
Linear Formula ZnCl2·xH2O
UN Number UN3260
Merck Index 14,10132
Solubility Information Soluble in water,ethanol (slightly),acetone (slightly),ether,and glycerol.
SMILES Cl[Zn]Cl
IUPAC Name dichlorozinc
Formula Weight 136.29 (Anhydrous)
Percent Purity 99.999%
Grade Puratronic
Odor Odorless
Assay (metals basis)
Chemical Name or Material Zinc chloride hydrate
Show More Show Less
missing translation for 'healthHazard1' H302-H314-H335
missing translation for 'healthHazard2' GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
missing translation for 'healthHazard3' P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 231-592-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.