Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Chloride, Purified, Anhydrous, Reagents

Catalog No. 01336866
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-866 500 g
01-336-868 100 g
01-336-867 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-336-866 Supplier Reagents Holdings Llc Supplier No. C2352000500C2
Add to Cart
Add to Cart

Zinc Chloride, Purified, Anhydrous, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7646-85-7
Molecular Formula Cl2Zn
Molecular Weight (g/mol) 136.28
InChI Key JIAARYAFYJHUJI-UHFFFAOYSA-L
Synonym Zinc Dichloride
IUPAC Name zinc(2+) dichloride
SMILES [Cl-].[Cl-].[Zn++]

Specifications

Boiling Point 732°C
Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 2.907
Chemical Name or Material Zinc Chloride, Purified, Anhydrous
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.