Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc ICP Standard, 10,000 ppm Zn in 5% HNO3, Ricca Chemical
SDP

Catalog No. PZN10KN500 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
500 mL
Packaging:
Natural LDPE Bottle
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
PZN10KN-500 500 mL Natural Poly Bottle
PZN10KN-50 50 mL Natural Poly Bottle
PZN10KN-100 100 mL Natural LDPE Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. PZN10KN-500 Supplier Ricca Chemical Company Supplier No. PZN10KN500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7440-66-6 , 7697-37-2 , 7732-18-5
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Packaging Natural Poly Bottle
Concentration or Composition Notes 10,000ppm Zn
CAS Min % 87.0, 10.42, 0.87
CAS Max % 90.0, 10.85, 0.9
Equivalence 1 mL = 10 mg Zn
Physical Form Liquid
Color Colorless
pH < 2
Melting Point Approximately 0°C
Type Standard
Quantity 500 mL
Chemical Name or Material Zinc ICP Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.