Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc(II) acetylacetonate, ca. 25% Zn, Acros Organics

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
14024-63-6
C10H14O4Zn
263.59
MFCD00000035
KUJHAYOLESEVSA-SUKNRPLKSA-L
zinc acetylacetonate hydrate,zinc 2,4-pentanedionate, hydrate,zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
131675103
zinc;(Z)-4-oxopent-2-en-2-olate;hydrate
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Zn+2]
14024-63-6
C10H14O4Zn
263.59
MFCD00000035
KUJHAYOLESEVSA-SUKNRPLKSA-L
zinc acetylacetonate hydrate,zinc 2,4-pentanedionate, hydrate,zinc 2+ bis 2z-4-oxopent-2-en-2-olate hydrate
131675103
zinc;(Z)-4-oxopent-2-en-2-olate;hydrate
CC(=CC(=O)C)[O-].CC(=CC(=O)C)[O-].O.[Zn+2]
Solubility Information Solubility in water: 6.9g/L (20°C)
Formula Weight 263.59
Physical Form Powder
Percent Purity ca. 25% Zn
Infrared Spectrum Authentic
Packaging Glass bottle
Solubility Solubility in water: 6.9g/L (20°C).
Boiling Point 129.0°C to 131.0°C (10.0mmHg)
Color White to Ivory
Melting Point 124.0°C to 126.0°C
Quantity 5g
Chemical Name or Material Zinc(II) acetylacetonate
Show More Show Less

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 237-860-3