Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Zinc, Metal Pieces
SCI_ED

Catalog No. S25644 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
100 Ea.
Packaging:
Pieces (1 x 1.2 x 0.05cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25644 100 Ea. Pieces (1 x 1.2 x 0.05cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S25644
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Reacts with water or moisture

This product is an Educational Grade Chemical — no C of A is available.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Density 7.14g/mL
Quantity 100 Ea.
Packaging Pieces (1 x 1.2 x 0.05cm)
Form Solid
Assay Percent Range 100% w/w
Linear Formula Zn
Formula Weight 65.38
Chemical Name or Material Zinc, Metal Pieces
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.