Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Metal Powder (Dust), Reagents

Catalog No. 01336870
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-870 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-870 Supplier Reagents Holdings Llc Supplier No. C2350500500A5
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zinc Metal Powder (Dust), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Silver-Gray
Boiling Point 907°C
Avg. Mol. Wt. or Mol. Wt. Range 65.38
Quantity 500 g
Packaging Poly Bottle
Form Solid
Chemical Name or Material Zinc Metal Powder (Dust)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.