Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Zinc nitrate hydrate

Catalog No. 230006250G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-000-6250G 250 g
23-000-625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-000-6250G Supplier Sigma Aldrich Supplier No. 230006250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Zinc nitrate hydrate

Specifications

CAS 13778-30-8
Melting Point 36°C
Density 2.065 g/mL (at 25°C)
Molecular Formula N2O6Zn -+ xH2O
Linear Formula Zn(NO3)2 -+ xH2O
Quantity 250 g
MDL Number MFCD00149888
Synonym Dinitric acid zinc salt hydrate
Molecular Weight (g/mol) 189.4
Percent Purity 99.999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.