Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc oxide, 99.5+%

Catalog No. AC194890050
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC194890050 5 kg Plastic Drum
AC194890010 1 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC194890050 Supplier Thermo Scientific Chemicals Supplier No. 194890050
Add to Cart

CAS: 1314-13-2 | OZn | 81.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Melting Point 1975.0°C
Color White or White-Yellow
Physical Form Amorphous Powder
Quantity 5 kg
Assay Percent Range 99.5% min. (Complexometry)
Linear Formula ZnO
Merck Index 15,10347
Solubility Information Solubility in water: 1.6 mg/l (29°C). Other solubilities: soluble in acids and alkalies, soluble in diluted acetic or mineral acids, ammoni, ammonium carbonate, fixed alkali hydroxide solutio, insoluble in alcohol
Formula Weight 81.38
Percent Purity ≥99.5%
Packaging Plastic Drum
Residue after Ignition 1% max. (1 g,500°C)
Specific Gravity 5.6
Density 5.6000g/mL
Chemical Name or Material Zinc oxide, 99.5+%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Avoid release to the environment.
EINECSNumber 215-222-5
RTECSNumber ZH4810000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.