Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc oxide, NanoTek™ ZN-2525, 40% in 1,2-propanediol monomethyl ether acetate, colloidal dispersion w/ dispersant, Thermo Scientific™

Catalog No. AA4564636
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4564636 500 g
AA4564622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4564636 Supplier Thermo Scientific Chemicals Supplier No. 4564636

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

  • Zinc oxide dispersed in Dowanol PMA suitable for incorporation in a wide variety of polar organic based coatings and formulations
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Color White
Physical Form Dry Powder
Quantity 500 g
Linear Formula ZnO
UN Number UN1993
Formula Weight 81.37
Concentration or Composition (by Analyte or Components) 40% in 1,2-propanediol monomethyl ether acetate
Odor Ester-like
Chemical Name or Material Zinc oxide, NanoTek™ ZN-2525, Colloidal dispersion with dispersant
Hazard Category Warning
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement GHS P Statement
P273-P501a
Avoid release to the environment.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.