Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc oxide sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.9% (metals basis)

Catalog No. AA41124KS
Click to view available options
Quantity:
1 Ea.

Catalog No. AA41124KS

Supplier: Thermo Scientific Chemicals 041124KS

Only null left
Add to Cart

CAS: 1314-13-2 | OZn | 81.38 g/mol

Zinc oxide is used as an additive in numerous materials and products including rubbers, plastics, ceramics, glass, cement, lubricants, paints, ointments, adhesives, sealants, pigments, foods, batteries, ferrites, fire retardants, and first-aid tapes. It is also used in oil, greases, ferrites, cosmetics, pharmaceuticals, medical, UV coatings, lacquers, ceramics and textiles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-13-2
Molecular Formula OZn
Molecular Weight (g/mol) 81.38
MDL Number MFCD00011300
InChI Key XLOMVQKBTHCTTD-UHFFFAOYSA-N
Synonym zinc oxide, zinc white, amalox, permanent white, Chinese white, snow white, emanay zinc oxide, felling zinc oxide, zinc oxide zno, akro-zinc bar 85
PubChem CID 14806
ChEBI CHEBI:36560
IUPAC Name oxozinc
SMILES O=[Zn]

Specifications

Quantity 1 Ea.
Solubility Information Soluble in diluted acid. Insoluble in water and alcohol.
Formula Weight 81.37
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Zinc oxide sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 215-222-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.