Learn More
Medchemexpress LLC Zinc phytate | 63903-51-5 | MFCD16879089 | 98.2% | 1040.22 | C6H6O24P6Zn6 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN258010MG
Encompass_Preferred
Zinc phytate is the zinc salt of inositol hexaphosphate used as a research reagent and precursor to inositol phosphates and pyrophosphates. It has reported activity attenuating Aβ oligomers and modulating autophagy-related proteins, and is used in studies of cardiovascular, metabolic, nervous system disorders, and cancer.
  • High reported purity: 98.2%.
  • Molecular formula C6H6O24P6Zn6 and molecular weight 1040.22.
  • Solid form; store sealed at 4°C away from moisture and light.
  • Serves as a precursor for inositol phosphate and pyrophosphate research.
  • Reported biological effects include attenuation of Aβ oligomers and upregulation of autophagy proteins.
  • Available in small research quantities suitable for laboratory experiments.
Catalog No. 50-002-89418