Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc powder, -100 mesh, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog No. p-4412898
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
5 x 1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA0042430 250 g
AA00424A1 1 kg
AA00424D9 5 x 1 kg
AA00424A7 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA0042430 Supplier Thermo Scientific Chemicals Supplier No. 00042430
Only null left
Add to Cart

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Gray
Density 7.14
Quantity 250 g
Mesh Size -100 Mesh
Sensitivity Air and moisture sensitive
Form Powder
Assay Percent Range 99.9% (metals basis)
Formula Weight 65.38
Chemical Name or Material Zinc
Hazard Category H250-H260-H500
Precautionary Statement P210-P222-P223-P231+P232-P280-P335+P334-P370+P378q-P501c
DOTInformation DOT Class: 4.3; Packaging Group: II
EINECSNumber 231-175-3
RTECSNumber ZG8600000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.