Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc (Powder/Certified), Fisher Chemical

Catalog No. Z5500 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
Z5-500 500 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. Z5-500

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 7440-66-6
Molecular Formula Zn
Molecular Weight (g/mol) 65.38
MDL Number MFCD00011291
InChI Key HCHKCACWOHOZIP-UHFFFAOYSA-N
Synonym dust, cinc, powder, merrillite, rheinzink, jasad, blue powder, granular, zinc, elemental, emanay dust
PubChem CID 23994
ChEBI CHEBI:30185
IUPAC Name zinc
SMILES [Zn]

Specifications

Color Blue
Melting Point 419°C
Boiling Point 908°C
Atomic Weight 65.37
Density 7.14g/cm³
Quantity 500 g
Identification Pass Test
Packaging Glass Bottle
Vapor Pressure 1mmHg at 487°C
Form Solid
Assay Percent Range ≥97 %
UN Number 1436
Formula Weight 65.41
Percent Purity ≥97%
Grade Certified
Chemical Name or Material Zinc
Show More Show Less
Safety handling Check Alert storage code Red
DOTInformation Class 4.3: Dangerous When Wet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.