Learn More
Zinc propionate, 97%
Description
Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used to reduce irritation from molds, fungi, and bacteria and as a topical antifungal agent.
Solubility
Soluble in alcohol, ether, chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform.
Notes
Hygroscopic, protect from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Quantity | 1000 g |
| Sensitivity | Hygroscopic |
| Merck Index | 14,10153 |
| Solubility Information | Soluble in alcohol,ether,chloroform. Miscible with ethanol; soluble in diethyl ether; slightly soluble in chloroform. |
| Formula Weight | 211.53 |
| Percent Purity | 97% |
| Chemical Name or Material | Zinc propionate |
Safety and Handling
| Hazard Category | H318 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P280-P305+P351+P338-P310 |
| EINECSNumber | 209-167-6 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.