Learn More
Cayman Chemical MIFhumnrecombInt 100ug
SDP
Supplier:  Cayman Chemical 14493100 UG
Encompass_Preferred
An inhibitor of SUR1/Kir6.2 SUR2A/Kir6.2 and SUR2B/Kir6.2 (IC50s 3 5.4 and 7.3 nM respectively in Xenopus oocytes expressing the mouse receptor) induces InsR complex dissociation and insulin degradation in HepG2 cells at 20 UM decreases blood glucose and plasma insulin levels in a KKAy mouse model of insulin-resistant type 2 diabetes at 0.05 mg/kg per day reduces HbA1c levels in the same model at 0.5 mg/kg per day
Catalog No. 50-296-9723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.