Learn More
Cayman Chemical ANTIBODY VUF10460 5mg
SDP
Supplier:  Cayman Chemical 146655 MG
Encompass_Preferred
An amino acid with antidiabetic activity increases glucose-stimulated insulin release from isolated rat islets and isolated perfused rat pancreas at 200 M increases PI3K activity in muscle and liver in rats at 18 mg/kg and in muscle in a rat model of type 2 diabetes induced by nicotinamide and STZ at 25 mg/kg decreases plasma glucose triglyceride LDL HDL and cholesterol levels in an STZ-induced rat model of type 1 diabetes at 50 mg/kg per day for four weeks
Catalog No. 50-296-9669
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.