Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Pyrithione, 48% Aqueous Suspension, Spectrum™ Chemical
SDP

Catalog No. 18607873 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
5 kg
12 kg
Packaging:
Amber Glass Bottle
Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-873 12 kg Poly Pail
18-607-870 100 g Amber Glass Bottle
18-607-871 500 g Amber Glass Bottle
18-607-872 2.5 kg Glass Bottle
18-607-875 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-873 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z102512KGBL
Add to Cart
Add to Cart

Z1025, 13463-41-7

Spectrum™ Chemical Zinc Pyrithione, 48 Percent Aqueous Suspension is a fungistatic and baceriostatic compound that has a variety of uses. It is used in outdoor paint and other products that provide protection against mildew and algae due to its low solubility in water (8 ppm at neutral pH). It is also often used in kitchen sponges as an antibacterial treatment. 48 Percent Aqueous Suspension.

Chemical Identifiers

CAS 13463-41-7
Molecular Formula C10H8N2O2S2Zn
Molecular Weight (g/mol) 317.69
InChI Key OTPSWLRZXRHDNX-UHFFFAOYSA-L
IUPAC Name zinc pyrithione
SMILES [Zn++].[O-][N+]1=CC=CC=C1[S-].[O-][N+]1=CC=CC=C1[S-]

Specifications

CAS Max % 1
Packaging Poly Pail
Quantity 12 kg
Percent Purity 48 to 52%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.