Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Stearate, Powder, USP, 12.5-14%, Spectrum™ Chemical
SDP

Catalog No. 18607877 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
Packaging:
Poly Pail
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-877 2.5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-877 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. ZI1152.5KG
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

ZI115, 557-05-1

Spectrum™ Chemical Zinc Stearate, Powder, USP is a mild antiseptic and astringent used as a local soothing application for inflammatory and irritating skin diseases and in pharmacuetical formulations as a lubricant in tablet and capsule manufacture. All Spectrum Chemical manufactured USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 557-05-1
Molecular Formula C36H70O4Zn
Molecular Weight (g/mol) 632.33
InChI Key XOOUIPVCVHRTMJ-UHFFFAOYSA-L
IUPAC Name zinc(2+) dioctadecanoate
SMILES [Zn++].CCCCCCCCCCCCCCCCCC([O-])=O.CCCCCCCCCCCCCCCCCC([O-])=O

Specifications

CAS Max % 1
Quantity 2.5 kg
Percent Purity 12.5 to 14%
Grade USP
Packaging Poly Pail
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.