Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfate concentrate, For 1L standard solution, 0.1 M ZnSO4, Solstice

Catalog No. 6001786 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-017-86 1 Ea.
60-046-058 6 x 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-017-86 Supplier Solstice Supplier No. 320471EA
Add to Cart
Add to Cart

for 1L standard solution, 0.1 M ZnSO4

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7733-02-0
Molecular Formula O4SZn
Molecular Weight (g/mol) 161.44
MDL Number MFCD00011302
InChI Key NWONKYPBYAMBJT-UHFFFAOYSA-L
Synonym zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
PubChem CID 24424
ChEBI CHEBI:35176
IUPAC Name zinc;sulfate
SMILES [Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 1 Ea.
Linear Formula ZnSO4
UN Number 3082
Formula Weight 161.45g/mol
Packaging Ampoule
Chemical Name or Material Zinc Sulfate Concentrate
Hazard Statement P273-P280-P305 + P351 + P338-P501
Precautionary Statement H318-H410
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.