Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfate heptahydrate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, 99.0-103.0%, Solstice

Catalog No. 60044863 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
4 x 5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-044-863 4 x 5 kg
60-020-16 1 kg
60-044-864 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-044-863 Supplier Solstice Supplier No. 144554X5KG

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, 99.0-103.0%

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2o
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Quantity 4 x 5 kg
Linear Formula ZnSO4 · 7H2O
UN Number UN3077
Formula Weight 287.56
Percent Purity 99.0 to 103.0%
Grade puriss.
Chemical Name or Material Zinc Sulfate Heptahydrate, Meets analytical specification of Ph. Eur.BP, USP, FCC
Hazard Statement P273-P280-P305 + P351 + P338-P501
Precautionary Statement H302-H318-H410
DOTInformation 9 / PGIII
EINECSNumber 231-793-3
RTECSNumber ZH5300000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.