Learn More
Specifications
Specifications
| Melting Point | 100°C |
| Color | White |
| pH | 4.4 to 6 |
| Physical Form | Solid |
| Quantity | 3 kg |
| Formula Weight | 287.56 |
| Percent Purity | 99.0 to 108.7% (USP) |
| Grade | FCC/USP |
| Identification | Pass Test (FCC), Pass Test (USP) |
| Packaging | Poly Bottle |
| Show More |
Safety and Handling
| Hazard Statement | WARNING! |
| Precautionary Statement | Emergency Overview Harmful if swallowed. May cause skin and respiratory tract irritation. Very toxic to aquatic organisms, may cause long-term adverse effects in the aquatic environment. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici. Get medical attention immediately if symptoms occur. NFPA Health:2 Flammability:0 Instability:0 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.