Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc Sulfate Monohydrate, Purified, Reagents

Catalog No. 01336863
Change view
Click to view available options
Quantity:
500 g
2.5 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-336-863 500 g
01-336-864 2.5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-336-863 Supplier Reagents Holdings Llc Supplier No. C2354000500A5
Add to Cart
Add to Cart

Zinc Sulfate Monohydrate, Purified, Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7446-19-7
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.45
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym Zinc Sulphate Monohydrate
IUPAC Name zinc(2+) hydrate sulfate
SMILES O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Color Colorless/White
Physical Form Solid
Quantity 500 g
Grade Pure
Packaging Poly Bottle
Specific Gravity 3.2
Chemical Name or Material Zinc Sulfate Monohydrate, Purified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.