Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zinc sulfide, 98%

Catalog No. AAA162960B
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA162960B 1000 g
AAA1629630 250 g
AAA162960I 5000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA162960B Supplier Thermo Scientific Chemicals Supplier No. A162960B
Add to Cart

CAS: 1314-98-3 | SZn | 97.44 g/mol

Zinc Sulfide is used as a friction material to carry sulfur, which hardens rotor surfaces and thereby reduces wear. Also in printing inks, UV-hardened systems, powder coatings, adhesives, insulating & sealing compounds, thermoplastic pigmentation, thermosets, flame resistant plastics, glass-fiber reinforced plastics, pigment concentrates, elastomers,. textile fibers, paper, mastics, lubricants, electroluminescent lamps, infrared windows, domes and optical elements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Zinc Sulfide is used as a friction material to carry sulfur, which hardens rotor surfaces and thereby reduces wear. Also in printing inks, UV-hardened systems, powder coatings, adhesives, insulating & sealing compounds, thermoplastic pigmentation, thermosets, flame resistant plastics, glass-fiber reinforced plastics, pigment concentrates, elastomers,. textile fibers, paper, mastics, lubricants, electroluminescent lamps, infrared windows, domes and optical elements.

Solubility
It is insoluble in water, soluble in acids.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Air and moisture sensitive. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-98-3
Molecular Formula SZn
Molecular Weight (g/mol) 97.44
MDL Number MFCD00011301
InChI Key DRDVZXDWVBGGMH-UHFFFAOYSA-N
Synonym zinc sulfide, sachtolith, sphalerite, zinc blende, albalith, cleartran, zinc monosulfide, pigment white 7, zinc sulfide zns, zinc sulphide
PubChem CID 14821
SMILES [S--].[Zn++]

Specifications

Melting Point 1700°C
Quantity 1000 g
Linear Formula ZnS
Merck Index 14,10160
Solubility Information It is insoluble in water,soluble in acids.
IUPAC Name sulfanylidenezinc
Formula Weight 97.43
Percent Purity 98%
Odor Odorless
Density 4.1
Chemical Name or Material Zinc sulfide
Show More Show Less
EINECSNumber 215-251-3
RTECSNumber ZH5400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.