Learn More
Supelco Inc ZINC VOLUMETRIC STANDARD 100G
Supplier:  Supelco Inc 1024090103
Encompass
Zinc volumetric standard, secondary reference material for complexometry, traceable to NIST SRM Certipur® Reag. Ph Eur. CAS No. 7440-66-6, EC Number 231-175-3.Made to order product. 6-8 weeks lead-time. Non returnable.
Catalog No. NC2736694
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.