Learn More
TARGETMOL CHEMICALS INC ZINC00640089 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T72968
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. ZINC00640089 is a selective lipid carrier protein-2 (LCN2) inhibitor. ZINC00640089 inhibited cell proliferation and cell viability and reduced AKT phosphorylation in SUM149 cells. ZINC00640089 can be used to study inflammatory breast cancer (IBC). purity: 99%
Catalog No. 50-257-9228
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.