Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Zinpyr-1, Fluorogenic Zn²+ Reporter, >95% Purity, 20 mg

Catalog No. 502583255
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-258-3255 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-258-3255 Supplier Abcam Supplier No. AB14534920MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cell-permeable fluorogenic Zn²+ reporter that induces positive fluorescence upon metal ion complexation.

  • Molecular weight: 823.72 g/mol
  • Purity: >95%
  • Product type: Cell-permeable fluorogenic Zn2+ reporter
  • Description: 2,7-Dichlorofluorescein (ab146332) derivative; acts as a PET sensor that induces positive fluorescence upon metal ion complexation
  • Constituents: 100% Zinpyr-1
  • Citation count: 8

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule Zinc (Zn2+)
Substrate Properties Chemical Substrate
Shipping Condition Ambient
Substrate Zn2+
Buffer 100% Zinpyr-1
Content And Storage Store at -20°C.
Detection Location In-Cell Detection
Kit Contents 20 mg Zinpyr-1
Detection Method Fluorescence
Formulation 100% Zinpyr-1
Quantity 20 mg
For Use With (Application) Fluorescence detection of Zn²+
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.