Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZIP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521066
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521066 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521066 Supplier Invitrogen™ Supplier No. PA521066
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

A suggested positive control is mouse kidney tissue lysate. PA5-21066 can be used with blocking peptide PEP-1180.

ZIP1, also known as ZIRTL (zinc-iron regulated transporter-like), is the first mammalian member of a family of divalent ion transporters. Zinc is an essential ion for cells and plays significant roles in the growth, development, and differentiation. ZIP1 expression is markedly downregulated in a number of cancerous tissues and is thought to function as a tumor suppressor gene in prostate cancer. More recent studies have shown that overexpression of ZIP1 and concomitant increased levels of intracellular zinc in PC-3 cells cause a significant inhibition of NF-kappa-B, leading to down-regulation of the antiapoptotic protein Bcl-2, Bcl-XL, and the apoptosis inhibitor XIAP, reducing the malignant potential of prostate cancer cells in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Slc39a1
Gene Accession No. Q9NY26, Q9QZ03
Gene Alias CGI-08; CGI-71; IRT1; mZIP1; SLC39A1; solute carrier family 39 (zinc transporter), member 1; solute carrier family 39 (zinc transporter), member 3; solute carrier family 39 member 1; Zinc transporter ZIP1; Zinc-iron-regulated transporter-like; Zip1; ZIP-1; ZIRTL; Zrt- and Irt-like protein 1; Zrt/Irt-like protein 1
Gene Symbols Slc39a1
Host Species Rabbit
Immunogen A 16 amino acid peptide near the amino terminus of human ZIP1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27173, 30791
Target Species Human, Mouse
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.