Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZIP10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557696 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557696 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557696 Supplier Thermo Scientific Supplier No. PA557696
Add to Cart
Edge
Add to Cart

ZIP10 Polyclonal Antibody

ZIP10 Polyclonal Antibody for ICC/IF, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZIP10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC39A10
Gene Accession No. Q9ULF5
Gene Alias 2900042E17Rik; 5430433I10; Kiaa1265; LZT-Hs2; mKIAA1265; Slc39a10; solute carrier family 39 (metal ion transporter), member 10; solute carrier family 39 (zinc transporter), member 10; solute carrier family 39 member 10; Zinc transporter ZIP10; ZIP10; ZIP-10; Zrt- and Irt-like protein 10
Gene Symbols SLC39A10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZIP10. Recombinant protein control fragment (Product #RP-102722).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57181
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.