Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) chloride, Reactor Grade, 99.5+% (metals basis), Thermo Scientific Chemicals

Catalog No. AA1210436
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
2 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1210436 500 g
AA12104A3 2 kg
AA1210409 10 g
AA1210422 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1210436 Supplier Thermo Scientific Chemicals Supplier No. 01210436

Zirconium tetrachloride is commonly utilized as a Lewis acid catalyst in many organic synthesis reactions. It is an important source for organometallic complexes of zirconium. It is practiced in many areas like nuclear industry, metal industry, photography, glass, and ceramic industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-11-6
Molecular Formula ZrCl4
MDL Number MFCD00011306
Synonym Zirconium tetrachloride

Specifications

Melting Point 331° (sublimation)
Physical Form Powder
Quantity 500 g
Assay Percent Range >99.5% (metals basis)
Solubility Information Soluble in alcohol,ether,hydrochloric acid.
Formula Weight 233.03
Grade Reactor
Sensitivity Moisture sensitive
Density 2.803 g/mL
Chemical Name or Material Zirconium(IV) chloride
Hazard Category H290-H302-H314
Precautionary Statement P234-P264b-P270-P280-P284-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P390-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 233-058-2
RTECSNumber ZH7175000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.