Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconium(IV) iodide, 99%, Thermo Scientific Chemicals

Catalog No. p-4904582
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1740014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA1740014 Supplier Thermo Scientific Chemicals Supplier No. 01740014
Only null left
Add to Cart

Light, Air and Moisture Sensitive

Used in the van Arkel-de Boer refining process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13986-26-0
Molecular Formula I4Zr
MDL Number MFCD00049641
Synonym zirconium iodide, unii-h6nzc1810v, zirconium iv ; iodide, acmc-1bzrf, zirconium 4+ tetraiodide

Specifications

Melting Point 431° (sublimation)
Color Yellow
Physical Form Powder
Quantity 25 g
Assay Percent Range 99%
Solubility Information Insoluble in water.
Formula Weight 598.84
Sensitivity Air, light, and moisture sensitive
Density 4.85
Chemical Name or Material Zirconium(IV) iodide
Hazard Category H314-H335
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 237-780-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.