Learn More
Description
- Chemical name: Zirconium Oxide
- Purity: ∼99%
- Molecular formula: ZrO2
- Molecular weight: 123.22
- Physical state: Solid
- Appearance: Powder crystals
- Color: White
- Odor: Odorless
- Melting point: 2856.0°C to 4919.0°C
- Boiling point: 4300°C
- Relative density: 5.68 to 5.85
- Solubility: Insoluble in water; slightly soluble in nitric acid
Specifications
Specifications
| Boiling Point | 4300°C |
| Melting Point | 2856.0°C to 4919.0°C |
| Color | White |
| CAS Max % | 1 |
| Physical Form | Solid |
| Quantity | 2.5 kg |
| Assay Percent Range | 99% |
| Molecular Formula | ZrO2 |
| Linear Formula | ZrO2 |
| Synonym | Zirconium Dioxide |
| Show More |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
