Alfa Aesar™ Zirconium powder, APS 2-3 micron

Manufacturer: Alfa Aesar™ 0084736

Quantity 500g

 View more versions of this product

Catalog No. AA0084736

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-67-7
    MDL Number MFCD00011303
    UN Number UN1358
    Chemical Name or Material Zirconium powder

    Specifications

    Health Hazard 3 GHS P Statement
    P210-P280-P240-P241-P370+P378b
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: Special powder for metal fires.
    Physical Form Powder
    MDL Number MFCD00011303
    Health Hazard 2 GHS H Statement
    H228
    Flammable solid.
    TSCA TSCA
    CAS 7440-67-7
    UN Number UN1358
    Health Hazard 1 Danger
    Storage Note 1
    • Packaged in water
    Odor Odorless
    Particle Size APS 2 to 3 Micron
    Color Dark gray
    DOT Information Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: ZIRCONIUM POWDER, WETTED
    RTECS Number ZH7070000
    Quantity 500g
    Chemical Name or Material Zirconium powder

    GHS H Statement
    H228
    Flammable solid.

    GHS P Statement
    P210-P280-P240-P241-P370+P378b
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Wear protective gloves/protective clothing/eye protection/face protection.
    Ground/bond container and receiving equipment.
    Use explosion-proof electrical/ventilating/lighting/ equipment.
    In case of fire: Use for extinction: Special powder for metal fires.

    • Packaged in water

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit