Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZKSCAN4 Monoclonal Antibody (OTI6G6), TrueMAB™, OriGene
SDP

Catalog No. p-200017777
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-8440 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-8440 Supplier OriGene Supplier No. TA800605
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

ZKSCAN4 may be involved in the transcriptional activation of MDM2 and EP300 genes.

Specifications

Antigen ZKSCAN4
Applications Western Blot
Classification Monoclonal
Clone OTI6G6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZKSCAN4
Gene Accession No. Q969J2
Gene Alias P1P373C6, ZNF307, ZNF427, ZSCAN36
Gene Symbols ZKSCAN4
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-316 of human ZKSCAN4 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387032
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.