Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZKSCAN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531512
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531512 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531512 Supplier Invitrogen™ Supplier No. PA531512
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: 293T, A431, Jurkat, Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

ZKSCAN4 may be involved in the transcriptional activation of MDM2 and EP300 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZKSCAN4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene ZKSCAN4
Gene Accession No. Q969J2
Gene Alias AI841875; Gm11272; OTTMUSG00000000421; P1P373C6; P373c6.1; p373c6.1 (novel C2H2 type zinc finger protein); Zfp307; Zinc finger protein 307; zinc finger protein 427; Zinc finger protein with KRAB and SCAN domains 4; zinc finger with KRAB and SCAN domains 4; ZKSCAN4; ZNF307; ZNF427; ZSCAN36
Gene Symbols ZKSCAN4
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ZNF307. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387032
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.