Learn More
CPC Scientific Z-Leu-Val-Gly-DMK 1MG
SDP
Supplier:  CPC Scientific SUBS042B
Encompass
SEQUENCE: Z-Leu-Val-Gly-DMKONE-LETTER SEQUENCE: ZLVG-DMKMOLECULAR FORMULA: C22H31N5O5MOLECULAR WEIGHT: 445.52STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [119670-30-3]SYNONYMS: Z-Leu-Val-Gly-diazomethylketoneRESEARCH AREA: Protein Kinases and PhosphatasesREFERENCES:SKU(s): SUBS-042A, SUBS-042B, SUBS-042C, SUBS-042D
Catalog No. 50-211-0525
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.