Learn More
Specifications
Specifications
| Gene ID (Entrez) | 10269 |
| Species | Human |
| Product Line | Novus |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB 100-2388 |
| Gene Alias | zona pellucida binding protein 2, zona pellucida-binding protein 2, ZPBP-like protein, ZPBPLMGC41930 |
| Gene Symbol | ZMPSTE24 |
| Quantity | 0.1 mg |
| Research Category | Angiogenesis, Cancer, Cell Biology, Cellular Markers, Signal Transduction, Ubiquitin Proteasome Pathway |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
