Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMPSTE24 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 01950193
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-950-193 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-950-193 Supplier Invitrogen™ Supplier No. PA116965
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Suggested positive control: human testis protein.

This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMPSTE24
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene ZMPSTE24
Gene Accession No. O75844, Q80W54
Gene Alias A530043O15Rik; CAAX prenyl protease 1 homolog; D030046F19; Face1; Face-1; Farnesylated proteins-converting enzyme 1; farnesylated-proteins converting enzyme 1; fi45a02; FLJ14968; HGPS; MADB; Prenyl protein-specific endoprotease 1; PRO1; STE24; Ste24p; wu:fi45a02; zgc:55655; zinc metallopeptidase STE24; zinc metallopeptidase STE24 homolog; zinc metallopeptidase, STE24; zinc metallopeptidase, STE24 homolog; zinc metalloproteinase (STE24 homolog, yeast); zinc metalloproteinase Ste24 homolog; zinc metalloproteinase, STE24 homolog; Zmpste24
Gene Symbols ZMPSTE24
Host Species Rabbit
Immunogen A synthetic peptide made to a C-terminal portion of the human ZMPSTE24 protein sequence (between residues 400-475).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10269, 230709
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.