Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZMYND11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5142033
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5142033 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5142033 Supplier Invitrogen™ Supplier No. PA5142033
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Additional Information: This antibody is expected to recognise all reported human isoforms (NP_006615.2; NP_997644.2; NP_001189393.1; NP_001189394.1; NP_001189395.1; NP_001189396.1; NP_001189397.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.

The protein encoded by this gene was first identified by its ability to bind the adenovirus E1A protein. The protein localizes to the nucleus. It functions as a transcriptional repressor, and expression of E1A inhibits this repression. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZMYND11
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene ZMYND11
Gene Accession No. Q15326
Gene Alias 2210402G22Rik; adenovirus 5 E1A-binding protein; Bone morphogenetic protein receptor-associated molecule 1; BRAM1; BS69; MGC111056; MRD30; Protein BS69; RP11-486H9.1; Unknown (protein for MGC:137432); zinc finger MYND domain-containing protein 11; Zinc finger MYND domain-containing protein 11-like protein; zinc finger MYND-type containing 11; zinc finger, MYND domain containing 11; zinc finger, MYND-type containing 11; ZMYND11
Gene Symbols ZMYND11
Host Species Goat
Immunogen Peptide with sequence SRVHGMHPKETT-C (aa 42-53).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10771
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.