Learn More
AdipoGen ZnAF-1 DA
SDP
Supplier:  AdipoGen CDXZ0007M005
Encompass_Preferred
Chemical. Formula C38H32N4O7. MW 656.68. Synthetic. Suitable for the fluorimetric detection of Zn2 after esterase hydrolysis. Spectral data lambdaex 492nm lambdaem 515nm in PBS.
Catalog No. 50-255-9886
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.