Learn More
AdipoGen ZnAF-1F DA
SDP
Supplier:  AdipoGen CDXZ0011M001
Encompass_Preferred
Chemical. Formula C38H30F2N4O7. MW 692.66. Synthetic. For application to living cells, the diacetyl derivative of ZnAF-1F, ZnAF-1F DA, was synthesized. ZnAF-1F DA can permeate through the cell membrane, and is hydrolyzed by esterase in the cytosol to yield ZnAF-1F, which is retained in the cells. ZnAF-1F DA has been used to measure the changes of intracellular Zn2 in cultured cells and hippocampal slices. Spectral data lambdaex 492nm lambdaem 514nm in PBS.
Catalog No. 50-255-9876
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.