Learn More
Enzo Life Sciences ZnAF-2 . tetrahydrochloride (1mg)
SDP
Supplier:  Enzo Life Sciences ALX620072M001
Encompass_Preferred
Non cell permeable fluorescent reagent (Ex(max): 492nm; Em(max): 514nm) for the detection of low concentration of zinc ion due to its strong affinity to zinc ion (dissociation constant: 2.7nM). The sample zinc ion can be specifically detected. Low background fluorescense supersensitizes the visualization for in vivo sample zinc ion. Also available as cell permeable derivative (ALX-620-076). ZnAF-2 conjugates TPEN (ALX-450-011) affinities with fluorescein and enhances the specificity for Zn2+. Alternative name: 6-[N-[N',N'-bis(2-Pyridinylmethyl)-2-aminoethyl]amino-3',6'-dihydroxy-spiro[isobenzofuran-1(3H),9'-[9H]xanthen]-3-one . 4 HCl. Formulation: Dissolved in 0.28ml DMSO. Purity: ≥98% (HPLC). Appearance: Pale yellow liquid. Long Term Storage: +4°C.
Catalog No. 50-200-9604
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.