Learn More
Abcam ZnAF-2F DA, Cell-Permeable Intracellular Zn2+ Probe, 1 mg
Description
- Molecular Weight: 692.66 g/mol
- Purity: >95%
- Cell Permeability: Cell-permeable diacetyl derivative of ZnAF-2F (ab145253)
- Mechanism: Hydrolyzed by esterase in the cytosol to ZnAF-2F, which is retained within the cell
- Applications: Used to measure intracellular Zn2+ changes in cultured cells and hippocampal slices
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Zinc (Zn2+) |
| Content And Storage | Store at -20°C. |
| Buffer | 100% ZnAF-2F DA |
| Detection Location | In-Cell Detection |
| Detection Method | Fluorescence |
| Formulation | 100% ZnAF-2F DA |
| Sample Type | Cultured cells, Hippocampal slices |
| Quantity | 1 mg |
| For Use With (Application) | Measuring intracellular Zn²+ in cultured cells and hippocampal slices |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.