Learn More
Abcam ZnAF-2F DA, Intracellular Zn2+ Probe, >95% Purity, 5 mg
Description
- Molecular weight: 692.66 g/mol
- Purity: >95%
- Description: Cell-permeable diacetyl derivative of ZnAF-2F (ab145253). Hydrolyzed by esterase in the cytosol to yield the metabolite ZnAF-2F, which is retained within the cell.
- Applications: Used to measure the changes of intracellular Zn2+ in cultured cells and hippocampal slices.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Zinc (Zn2+) |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% ZnAF-2F DA |
| Content And Storage | Store at -20°C. |
| Detection Location | In-Cell Detection |
| Detection Method | Fluorescence |
| Sample Type | Cultured cells, Hippocampal slices |
| Quantity | 5 mg |
| For Use With (Application) | Measuring changes of intracellular Zn2+ in cultured cells and hippocampal slices |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.