Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF10 Antibody (CL9747) [DyLight 680], R&D Systems™
SDP

Catalog No. NBP344233FR
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP344233FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP344233FR Supplier R&D Systems Supplier No. NBP344233FR

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

ZNF10 Monoclonal specifically detects ZNF10 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Western Blot.

Specifications

Antigen ZNF10
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone CL9747
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias KOX1zinc finger protein 10 (KOX 1), zinc finger protein 10, Zinc finger protein KOX1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P21506, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 7556
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.