Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF101 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554804 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554804 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554804 Supplier Invitrogen™ Supplier No. PA554804
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Immunogen sequence: GKVFLRHSFL DRHMRAHAGH KRSECGGEWR ETPRKQKQHG KASISPSSGA RRTVTPTRKR PYECKVCGKA FNSPNLFQIH Q Highest antigen sequence identity to the following orthologs: Mouse - 43%, Rat - 43%.

Zinc finger proteins (ZNFs), such as ZNF101, bind nucleic acids and perform many key functions, the most important of which is regulating transcription (summary by Bellefroid et al., 1993 ). See ZNF91 for general information on ZNFs.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF101
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ZNF101
Gene Accession No. Q8IZC7
Gene Alias DKFZp570I0164; HZF12; MGC149565; MGC149566; zinc finger protein 101; zinc finger protein 101 (Y2); zinc finger protein 12; zinc finger protein HZF12; ZNF101
Gene Symbols ZNF101
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ZNF101. Recombinant protein control fragment (Product #RP-93630).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94039
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.